siRNA Details
Gene Symbol: N4bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030563.2 | NP_085040.2 | 5 | 2407 | ||
GenBank mRNA | AK172981.1 | - | 5 | 2389 | ||
BC004022.1 | - | 2 | 954 | |||
BC089324.1 | - | 4 | 1572 | |||
BC111031.1 | - | 2 | 852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
