siRNA Details
Gene Symbol: Fcrls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030707.3 | NP_109632.2 | 4 | 536 | ||
GenBank mRNA | AB028174.1 | - | 4 | 567 | ||
AF302046.1 | - | 4 | 605 | |||
AF329486.1 | - | 4 | 537 | |||
AK034125.1 | - | 4 | 537 | |||
AK076123.1 | - | 4 | 570 | |||
AY506556.1 | - | 4 | 576 | |||
AY506557.1 | - | 4 | 576 | |||
BC016551.1 | - | 4 | 579 | 3602803 |
