siRNA Details
Gene Symbol: Nlrp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031389.2 | NP_113566.2 | 8 | 2723 | ||
GenBank mRNA | AF285581.1 | - | 6 | 1991 | ||
AK133284.1 | - | 8 | 2725 | |||
AK166817.1 | - | 8 | 2704 | |||
AK166869.1 | - | 8 | 2949 | |||
AY596194.1 | - | 8 | 2723 | |||
BC120833.1 | - | 6 | 1984 | 40129756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
