siRNA Details
Gene Symbol: Cep41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ278891.1 | - | 9 | 774 | ||
AK006059.1 | - | 9 | 762 | |||
AK132495.1 | - | 9 | 763 | |||
AK161461.1 | - | 9 | 748 | |||
AK166974.1 | - | 10 | 891 | |||
AK170213.1 | - | 9 | 787 | |||
BC031892.1 | - | 9 | 744 | 5365552 |
