siRNA Details
Gene Symbol: PITPNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006224.3 | NP_006215.1 | 3 | 427 | ||
XM_011523939.1 | XP_011522241.1 | 3 | 414 | |||
XM_011523940.1 | XP_011522242.1 | 3 | 849 | |||
GenBank mRNA | AL542612.3 | - | 3 | 425 | ||
BC027177.1 | - | 3 | 357 | |||
BC068538.1 | - | 3 | 310 | |||
BC082976.1 | - | 3 | 342 | 6578454 | ||
D30036.1 | - | 3 | 379 | |||
M73704.1 | - | 3 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
