siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 5, 6 | 1525 | ||
GenBank mRNA | AF330197.1 | - | 5, 6 | 1521 | ||
AK036351.1 | - | 4, 5 | 1260 | |||
AK048110.1 | - | 4, 5 | 1291 | |||
AK137148.1 | - | 3, 4 | 863 | |||
AK169999.1 | - | 4, 5 | 1635 | |||
AK171932.1 | - | 4, 5 | 1463 | |||
BC069835.1 | - | 5, 6 | 1381 | 30094289 |
