siRNA Details
Gene Symbol: PRKAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006253.4 | NP_006244.2 | 1, 2 | 456 | ||
XM_005253909.1 | XP_005253966.1 | 2, 3 | 358 | |||
GenBank mRNA | AF022116.1 | - | 1, 2 | 432 | ||
AJ224515.1 | - | 1, 2 | 167 | |||
AK301165.1 | - | 2, 3 | 305 | |||
AK312495.1 | - | 2, 3 | 270 | |||
BC001007.2 | - | 1, 2 | 433 | 2961281 | ||
BC001056.2 | - | 1, 2 | 433 | 2961281 | ||
BC001823.2 | - | 1, 2 | 433 | 2961281 | ||
BC017671.1 | - | 1, 2 | 442 | 3884583 | ||
BC018818.2 | - | 1, 2 | 433 | |||
BU539177.1 | - | 1, 2 | 457 | |||
Y12556.1 | - | 1, 2 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
