siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164271.1 | NP_001157743.1 | 9, 10 | 2499 | ||
NM_001316668.1 | NP_001303597.1 | 9, 10 | 2687 | |||
NM_006094.4 | NP_006085.2 | 9, 10 | 2757 | |||
NM_182643.2 | NP_872584.2 | 13, 14 | 4192 | |||
XM_005273374.1 | XP_005273431.1 | 13, 14 | 4073 | |||
XM_017012951.1 | XP_016868440.1 | 13, 14 | 3943 | |||
XM_017012952.1 | XP_016868441.1 | 9, 10 | 2490 | |||
GenBank mRNA | AB051510.1 | - | 13, 14 | 4078 | ||
AF026219.1 | - | 9, 10 | 2710 | |||
AF035119.1 | - | 9, 10 | 2761 | |||
AK025544.1 | - | 9, 10 | 2686 | |||
AK025733.1 | - | 4, 5 | 933 | |||
AK289734.1 | - | 9, 10 | 2680 | |||
AK291682.1 | - | 13, 14 | 4033 | |||
AK299049.1 | - | 9, 10 | 2499 | |||
BC054511.1 | - | 13, 14 | 3947 | 5240161 |

More Information
Other Species Targeted

Gene Ontology Categories:
