siRNA Details
Gene Symbol: ST3GAL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271142.1 | NP_001258071.1 | 10 | 1164 | ||
NM_001271145.1 | NP_001258074.1 | 9 | 1105 | |||
NM_001271146.1 | NP_001258075.1 | 9 | 1094 | |||
NM_001271147.1 | NP_001258076.1 | 9 | 1068 | |||
NM_001271148.1 | NP_001258077.1 | 6 | 852 | |||
NM_001323352.1 | NP_001310281.1 | 10 | 1266 | |||
NM_001323353.1 | NP_001310282.1 | 8 | 1030 | |||
NM_001323358.1 | NP_001310287.1 | 11 | 1275 | |||
NM_001323359.1 | NP_001310288.1 | 10 | 1175 | |||
NM_001323360.1 | NP_001310289.1 | 11 | 1336 | |||
NM_001323362.1 | NP_001310291.1 | 9 | 1202 | |||
NM_001323363.1 | NP_001310292.1 | 8 | 1041 | |||
NM_001323364.1 | NP_001310293.1 | 9 | 1213 | |||
NM_001323365.1 | NP_001310294.1 | 10 | 1105 | |||
NM_001323366.1 | NP_001310295.1 | 11 | 1175 | |||
NM_001323367.1 | NP_001310296.1 | 9 | 922 | |||
NM_001323368.1 | NP_001310297.1 | 9 | 930 | |||
NM_006100.3 | NP_006091.1 | 10 | 1277 | |||
GenBank mRNA | AB022918.1 | - | 9 | 1044 | ||
AF119391.1 | - | 9 | 1088 | |||
AK001922.1 | - | 9 | 855 | |||
AK315111.1 | - | 10 | 1268 | |||
AL539506.3 | - | 6 | 709 | |||
BC001266.2 | - | 10 | 1092 | |||
BC023312.1 | - | 10 | 1253 | 4996181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
