siRNA Details
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080968.1 | NP_001074437.1 | 19 | 1991 | ||
NM_133852.2 | NP_598613.2 | 20 | 2072 | |||
GenBank mRNA | AK076370.1 | - | 14 | 1690 | ||
AK077516.1 | - | 19 | 1996 | |||
AK220509.1 | - | 20 | 2000 | |||
BC011407.1 | - | 18 | 1792 | 3596079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
