siRNA Details
Gene Symbol: Mdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081392.1 | NP_001074861.1 | 97 | 16229 | ||
GenBank mRNA | AK142906.1 | - | 3 | 332 | ||
BC085230.1 | - | 5 | 579 |

More Information
Other Species Targeted

Gene Ontology Categories:
