siRNA Details
Gene Symbol: Mdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081392.1 | NP_001074861.1 | 98, 99 | 16378 | ||
GenBank mRNA | BC085230.1 | - | 6, 7 | 728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
