siRNA Details
Gene Symbol: Chpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133913.2 | NP_598674.2 | 3 | 2288 | ||
GenBank mRNA | AK089404.1 | - | 3 | 2298 | ||
AK122504.1 | - | 2 | 2935 | |||
AK138814.1 | - | 2294 | ||||
AK219661.1 | - | 2 | 220 | |||
BC019714.1 | - | 2 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
