siRNA Details
Gene Symbol: Ttll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142732.1 | NP_001136204.1 | 4 | 569 | ||
NM_133923.6 | NP_598684.4 | 4 | 686 | |||
GenBank mRNA | AK029462.1 | - | 4 | 570 | ||
AM690746.1 | - | 4 | 686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
