siRNA Details
Gene Symbol: Ttll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142732.1 | NP_001136204.1 | 6 | 1286 | ||
GenBank mRNA | AK029462.1 | - | 6 | 1287 | ||
BC021404.1 | - | 5 | 1035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
