siRNA Details
Gene Symbol: Nlrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133946.2 | NP_598707.2 | 4 | 2048 | ||
GenBank mRNA | AK039531.1 | - | 1 | 1332 | ||
AY355343.1 | - | 1 | 986 | |||
BC013519.1 | - | 1 | 955 | |||
BC031139.1 | - | 1 | 1040 | 4972485 | ||
FJ222825.1 | - | 4 | 1994 | |||
JF810536.1 | - | 3 | 2433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
