siRNA Details
Gene Symbol: Fbxl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028149.1 | NP_082425.1 | 15 | 8368 | ||
GenBank mRNA | AK028535.1 | - | 1 | 3185 | ||
BC004087.1 | - | 1 | 1375 | |||
CX211402.1 | - | 1 | 62 |
