siRNA Details
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110240.1 | NP_001103710.1 | 10 | 2767 | ||
NM_172426.2 | NP_766014.1 | 11 | 2857 | |||
GenBank mRNA | AK083127.1 | - | 11 | 2741 | ||
BC115867.1 | - | 10 | 2769 | 40081382 | ||
BC116637.1 | - | 9 | 2583 | 40081381 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031743, AF021923.
Gene Ontology Categories:
