siRNA Details
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 13, 14 | 2231 | ||
NM_001109686.2 | NP_001103156.1 | 12, 13 | 2072 | |||
NM_001109687.2 | NP_001103157.1 | 12, 13 | 2087 | |||
NM_172545.4 | NP_766133.2 | 13, 14 | 2210 | |||
GenBank mRNA | AB205007.1 | - | 13, 14 | 2237 | ||
AK049454.1 | - | 10, 11 | 1481 | |||
AK052174.1 | - | 13, 14 | 2194 | |||
AK054555.1 | - | 13, 14 | 2179 | |||
AK082062.1 | - | 13, 14 | 2221 | |||
BC056938.1 | - | 10, 11 | 1788 | |||
BC089302.1 | - | 10, 11 | 1940 |
