siRNA Details
Gene Symbol: 2900052L18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013668.1 | - | 2 | 787 | ||
AK034741.1 | - | 1 | 872 | |||
AK162948.1 | - | 1 | 973 |
Gene Symbol: LOC102640779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094254.1 | - | 1 | 645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
