siRNA Details
Gene Symbol: Nav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111016.1 | NP_001104486.1 | 5 | 964 | ||
NM_175272.3 | NP_780481.2 | 4 | 1226 | |||
GenBank mRNA | AK031334.1 | - | 4 | 524 | ||
AK039374.1 | - | 4 | 1234 | |||
AK040152.1 | - | 3 | 511 | |||
AK084063.1 | - | 5 | 942 | |||
AK129480.1 | - | 3 | 452 | |||
AK158337.1 | - | 4 | 638 | |||
BC150679.1 | - | 3 | 214 |
