siRNA Details
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175275.4 | NP_780484.2 | 23, 24 | 3989 | ||
GenBank mRNA | AF483477.1 | - | 6, 7 | 852 | ||
AK029118.1 | - | 9, 10 | 1518 | |||
AK040554.1 | - | 1, 2 | 136 | |||
BC027370.1 | - | 3, 4 | 704 | |||
BC089374.1 | - | 9, 10 | 1692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
