siRNA Details
Gene Symbol: Fbxo43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081253.1 | NP_001074722.1 | 2 | 956 | ||
GenBank mRNA | AK019714.1 | - | 2 | 892 | ||
AK030012.1 | - | 2 | 780 | |||
BC098484.1 | - | 2 | 956 | 30053208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
