siRNA Details
Gene Symbol: Plekhg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081333.1 | NP_001074802.1 | 12 | 2076 | ||
GenBank mRNA | AK029859.1 | - | 10 | 2748 | ||
AK131125.1 | - | 10 | 2354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
