siRNA Details
Gene Symbol: Pus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK173290.1 | - | 12 | 1619 | ||
BC008544.1 | - | 12 | 1633 | 3497634 | ||
BC138533.1 | - | 12 | 1521 | 8861553 | ||
BC145316.1 | - | 13 | 1539 | 9053871 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
