siRNA Details
Gene Symbol: Pus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK173290.1 | - | 13, 14 | 1797 | ||
AK180496.1 | - | 1, 2 | 75 | |||
BC008544.1 | - | 13, 14 | 1811 | 3497634 | ||
BC138533.1 | - | 13, 14 | 1699 | 8861553 | ||
BC145316.1 | - | 14, 15 | 1717 | 9053871 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
