siRNA Details
Gene Symbol: Pus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289780.1 | NP_001276709.1 | 17 | 3001 | ||
NM_001289781.1 | NP_001276710.1 | 16 | 3334 | |||
NM_178403.5 | NP_848490.2 | 16 | 2983 | |||
GenBank mRNA | AK145380.1 | - | 16 | 3333 | ||
AK160849.1 | - | 16 | 2935 | |||
AK173290.1 | - | 16 | 2922 | |||
BC008544.1 | - | 16 | 2936 | 3497634 | ||
BC010600.1 | - | 8 | 1718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
