siRNA Details
Gene Symbol: Chsy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081328.1 | NP_001074797.1 | 1 | 1092 | ||
GenBank mRNA | AK019528.1 | - | 1 | 79 | ||
AK038429.1 | - | 1 | 1033 | |||
AK043732.1 | - | 1 | 1092 | |||
AK044099.1 | - | 1 | 1091 | |||
AK048196.1 | - | 1 | 1004 | |||
AK220519.1 | - | 979 | ||||
BC151164.1 | - | 1 | 782 | 9088053 | ||
BC151165.1 | - | 1 | 782 | 9088054 | ||
BC157985.1 | - | 1 | 782 | 9088141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
