siRNA Details
Gene Symbol: Zscan29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290819.1 | NP_001277748.1 | 1 | 1198 | ||
NM_001290820.1 | NP_001277749.1 | 1 | 1198 | |||
GenBank mRNA | AK031943.1 | - | 1 | 1181 | ||
AK050477.1 | - | 1 | 1198 | |||
BC031552.1 | - | 1 | 315 | |||
BC103777.1 | - | 1 | 309 |

More Information
Other Species Targeted

Gene Ontology Categories:
