siRNA Details
Gene Symbol: Zscan29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290820.1 | NP_001277749.1 | 2 | 2071 | ||
GenBank mRNA | AK031943.1 | - | 2 | 2054 | ||
AK050477.1 | - | 2 | 2071 | |||
BC031552.1 | - | 2 | 1188 | |||
BC103777.1 | - | 2 | 1182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
