siRNA Details
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178897.4 | NP_849228.1 | 9 | 4392 | ||
GenBank mRNA | AK045279.1 | - | 9 | 4377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
