siRNA Details
Gene Symbol: Prmt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081240.3 | NP_001074709.1 | 1, 2 | 342 | ||
GenBank mRNA | AK045296.1 | - | 1, 2 | 333 | ||
AK045315.1 | - | 1, 2 | 332 | |||
AK154546.1 | - | 1, 2 | 286 | |||
AK161023.1 | - | 1, 2 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
