siRNA Details
Gene Symbol: Tanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181071.3 | NP_851416.2 | 4 | 453 | ||
GenBank mRNA | AK051973.1 | - | 4 | 449 | ||
AK052263.1 | - | 3 | 186 | |||
BP766228.1 | - | 4 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
