siRNA Details
Gene Symbol: Ipw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019361.1 | - | 694 |
Gene Symbol: Anks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128086.2 | NP_001121558.2 | 17, 18 | 2919 | ||
NM_001177396.1 | NP_001170867.1 | 3, 4 | 750 | |||
NM_001177397.1 | NP_001170868.1 | 3, 4 | 365 | |||
NM_001177398.1 | NP_001170869.1 | 3, 4 | 750 | |||
NM_181398.3 | NP_852063.1 | 3, 4 | 750 | |||
GenBank mRNA | AK032211.1 | - | 3, 4 | 752 | ||
AK034154.1 | - | 4, 5 | 385 | |||
AK049899.1 | - | 3, 4 | 695 | |||
AK132228.1 | - | 3, 4 | 717 | |||
AK163750.1 | - | 3, 4 | 724 | |||
BC098373.1 | - | 3, 4 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
