siRNA Details
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198102.2 | NP_932770.2 | 8 | 1183 | ||
GenBank mRNA | AK089980.1 | - | 9 | 1164 | ||
AK164994.1 | - | 3 | 324 | |||
AK171708.1 | - | 7 | 2403 | |||
BC023018.1 | - | 5 | 563 | |||
BC057448.1 | - | 8 | 1138 | 4016620 | ||
BC058764.1 | - | 9 | 1448 |
