siRNA Details
Gene Symbol: Atf7ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019426.2 | NP_062299.2 | 2 | 1736 | ||
GenBank mRNA | AK132359.1 | - | 1 | 356 | ||
AK134252.1 | - | 2 | 1743 | |||
AK167542.1 | - | 3 | 1782 | |||
BC037617.1 | - | 1738 | ||||
BC052460.1 | - | 2 | 1736 | 6834550 | ||
BC060246.1 | - | 2 | 1719 | 5705945 |
