siRNA Details
Gene Symbol: Plce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019588.2 | NP_062534.2 | 1 | 1008 | ||
GenBank mRNA | AB076247.1 | - | 1 | 1008 | ||
BC025027.1 | - | 1 | 747 | |||
BC138349.1 | - | 1 | 996 | 8861369 | ||
BC138350.1 | - | 1 | 996 | 8861370 | ||
BC145246.1 | - | 1 | 996 | 9053798 |
