siRNA Details
Gene Symbol: Ercc6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013608.2 | NP_001013626.2 | 2 | 434 | ||
NM_023507.3 | NP_075996.2 | 2 | 420 | |||
GenBank mRNA | AF217319.1 | - | 1 | 237 | ||
AK002307.1 | - | 2 | 421 | |||
AK035054.1 | - | 2 | 386 | |||
AK049874.1 | - | 2 | 746 | |||
AK050437.1 | - | 1 | 270 | |||
BU148612.1 | - | 2 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
