siRNA Details
Gene Symbol: Ercc6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013608.2 | NP_001013626.2 | 2, 3 | 631 | ||
NM_023507.3 | NP_075996.2 | 2, 3 | 617 | |||
GenBank mRNA | AF217319.1 | - | 1, 2 | 434 | ||
AK002307.1 | - | 2, 3 | 618 | |||
AK035054.1 | - | 2, 3 | 583 | |||
AK049874.1 | - | 2, 3 | 943 | |||
BU148612.1 | - | 2, 3 | 649 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006253498.
Gene Ontology Categories:
