siRNA Details
Gene Symbol: 4930550L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023774.3 | NP_076263.1 | 2 | 509 | ||
GenBank mRNA | AF319984.1 | - | 1 | 359 | ||
AK016090.1 | - | 2 | 487 | |||
BC087920.1 | - | 2 | 509 | 30281980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
