siRNA Details
Gene Symbol: CLEC10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006344.2 | NP_006335.2 | 5, 6 | 659 | ||
XM_005256412.1 | XP_005256469.1 | 5, 6 | 685 | |||
GenBank mRNA | AK291975.1 | - | 5, 6 | 622 | ||
AK292363.1 | - | 5, 6 | 626 | |||
AL546075.3 | - | 5, 6 | 608 | |||
BC027858.1 | - | 5, 6 | 606 | 5225381 | ||
BI910082.1 | - | 5, 6 | 655 | |||
D50532.1 | - | 5, 6 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
