siRNA Details
Gene Symbol: Ak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027592.3 | NP_081868.1 | 5 | 528 | ||
GenBank mRNA | AK012905.1 | - | 5 | 478 | ||
AK014903.1 | - | 4 | 450 | |||
BC019453.1 | - | 5 | 479 | 4024616 | ||
BG084768.2 | - | 3 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
