siRNA Details
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099637.2 | NP_001093107.2 | 16 | 4323 | ||
GenBank mRNA | AK016937.1 | - | 4 | 584 | ||
AK038878.1 | - | 3 | 892 | |||
AK172953.1 | - | 4 | 1832 | |||
BC038377.1 | - | 4 | 1784 | |||
BC085106.1 | - | 4 | 569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
