siRNA Details
Gene Symbol: Lrrc72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027699.2 | NP_081975.1 | 5 | 398 | ||
GenBank mRNA | AK007140.1 | - | 5 | 399 | ||
BC107290.1 | - | 4 | 304 | 40053043 | ||
BC107291.1 | - | 4 | 304 | 40053045 |
