siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 8 | 1200 | ||
GenBank mRNA | AK011615.1 | - | 8 | 710 | ||
AK018573.1 | - | 8 | 740 | |||
AK050302.1 | - | 8 | 732 | |||
AK136140.1 | - | 8 | 740 | |||
BC054720.1 | - | 8 | 718 | 5689248 |

More Information
Other Species Targeted

Gene Ontology Categories:
