siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 11 | 1388 | ||
GenBank mRNA | AK018573.1 | - | 11 | 928 | ||
AK050302.1 | - | 11 | 920 | |||
AK136140.1 | - | 11 | 928 | |||
AK200331.1 | - | 2 | 39 | |||
BC043674.1 | - | 4 | 365 | 5044565 | ||
BC054720.1 | - | 11 | 906 | 5689248 |

More Information
Other Species Targeted

Gene Ontology Categories:
