siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 28 | 3000 | ||
GenBank mRNA | AK050302.1 | - | 28 | 2532 | ||
AK135468.1 | - | 4 | 473 | |||
BC007154.1 | - | 922 | ||||
BC025808.1 | - | 13 | 1187 | |||
BC038055.1 | - | 2 | 267 | |||
BC043674.1 | - | 21 | 1977 | 5044565 | ||
BC054720.1 | - | 28 | 2518 | 5689248 |

More Information
Other Species Targeted

Gene Ontology Categories:
