siRNA Details
Gene Symbol: Ubiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027873.2 | NP_082149.1 | 2 | 1772 | ||
GenBank mRNA | AK004550.1 | - | 2 | 1772 | ||
AK078442.2 | - | 2 | 1773 | |||
BC015303.1 | - | 2 | 1480 | 4241204 |
