siRNA Details
Gene Symbol: NEBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006393.2 | NP_006384.1 | 1, 2 | 486 | ||
XM_005252342.4 | XP_005252399.1 | 1, 2 | 590 | |||
XM_005252343.4 | XP_005252400.1 | 1, 2 | 590 | |||
XM_005252344.4 | XP_005252401.1 | 1, 2 | 590 | |||
XM_017015469.1 | XP_016870958.1 | 1, 2 | 590 | |||
GenBank mRNA | BC110452.1 | - | 1, 2 | 173 | 40027448 | |
BC110453.1 | - | 1, 2 | 173 | 40027449 | ||
Y16241.1 | - | 1, 2 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
