siRNA Details
Gene Symbol: AGPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006412.3 | NP_006403.2 | 3, 4 | 595 | ||
GenBank mRNA | AF000237.1 | - | 3, 4 | 559 | ||
AF011374.1 | - | 3, 4 | 553 | |||
BC000026.2 | - | 3, 4 | 538 | 3507150 | ||
BC007269.1 | - | 1, 2 | 170 | |||
BC019292.1 | - | 3, 4 | 547 | 2823580 | ||
BF347518.1 | - | 3, 4 | 596 | |||
U56418.2 | - | 3, 4 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
